Sodium glucoheptonate

Supplier: Thermo Scientific Chemicals

H36703-22 H36703-36 H36703-A1
AAH36703-22EA 50.4 CAD
AAH36703-22 AAH36703-36 AAH36703-A1
Sodium glucoheptonate
Sodium glucoheptonate
Storage Temperature: Refrigerator
MDL Number: MFCD00013425
CAS Number: 13007-85-7
EINECS: 235-849-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR