Cephalothin

Supplier: Thermo Scientific Chemicals
Danger

H36175-06 H36175-14
AAH36175-06EA 144.41 CAD
AAH36175-06 AAH36175-14
Cephalothin
Cephalothin
Formula: C₁₆H₁₆N₂O₆S₂
Melting Pt: 160…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00242614
CAS Number: 153-61-7
EINECS: 205-815-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR