Diloxanide (furoate)

Supplier: Thermo Scientific Chemicals
Warning

H36096-03 H36096-MD
AAH36096-03EA 857.94 CAD
AAH36096-03 AAH36096-MD
Diloxanide (furoate)
Diloxanide (furoate)
Formula: C₁₄H₁₁Cl₂NO₄
Storage Temperature: Ambient
MDL Number: MFCD00083302
CAS Number: 3736-81-0
EINECS: 223-108-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR