Ethylboronic acid pinacol ester 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethane, Ethylboronic acid Pinacol ester

H36868-03 H36868-06 H36868-14
AAH36868-03EA 59.25 CAD
AAH36868-03 AAH36868-06 AAH36868-14
Ethylboronic acid pinacol ester 99%
Ethylboronic acid pinacol ester
Formula: C₈H₁₇BO₂
MW: 156.03 g/mol
Boiling Pt: 79…81°C / 120mbar
Storage Temperature: Ambient
MDL Number: MFCD19441153
CAS Number: 82954-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR