Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trityltetra(pentafluorophenyl)borate, Triphenylmethylium tetrakis(perfluorophenyl)borate, Trityl tetrakis(pentafluorophenyl)borate, Triphenylcarbenium Tetrakis(pentafluorophenyl)borate, Tritylium Tetrakis(pentafluorophenyl)borate

H36154-03 H36154-06
AAH36154-03EA 287.56 CAD
AAH36154-03 AAH36154-06
Triphenylmethylium tetrakis(pentafluorophenyl)borate 97%
Triphenylmethylium tetrakis(pentafluorophenyl)borate
Formula: C₄₃H₁₅BF₂₀
MW: 922.37 g/mol
Melting Pt: 180…185°C
Storage Temperature: Ambient
MDL Number: MFCD03426981
CAS Number: 136040-19-2
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR