VWR® L-Shape Cell Spreader, Sterile
Supplier: VWR International
76207-748 76208-438
76207-748CS 243.45 CAD
76207-748 76208-438
VWR® L-Shape Cell Spreader, Sterile
Spreaders Cell Spreaders
Disposable L-shaped cell spreaders are ideal for spreading cell or bacterial cultures evenly.
  • L-shape allows application on the entire dish or plate surface
  • Smooth spreading surface with slight upward return reduces the risk of damage to samples
  • Sterilized by electron beam irradiation
  • Non-pyrogenic
Spreaders made of polypropylene and pre-sterilized, eliminating the need for flaming or autoclaving.
Certifications: Manufactured in a class 100000 cleanroom environment; ISO13485:2003 and ISO 9001:2008.
Packaging: Individual wrap and bulk packaging available.
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M