Multiple Affinity Removal Column HSA/IgG, Agilent Technologies
Supplier: Agilent Technologies
5188-8826 5188-8826
97066-658EA 3288.17 CAD
97066-658 CA97066-658
Multiple Affinity Removal Column HSA/IgG, Agilent Technologies
Chromatography Columns
The multiple affinity removal LC column – human albumin/IgG is designed to chromatographically remove the two most interfering high-abundant proteins (albumin and IgG) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
  • Serum injection volume is 100 µl per run for the 4.6 mm id column
  • Optimize sample recovery by utilizing rapid, single injection protocols and optimized buffers
  • Column is compatible with most FPLC or HPLC pumps or instruments
Removal of albumin and IgG improves the subsequent LC/MS and electrophoretic analysis ofMore Product Information