XL10-Gold Ultracompetent Cells
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
200314 200315
CAST200314EA 258.42 CAD
CAST200314 CAST200315 CAST200317
XL10-Gold Ultracompetent Cells
Competent Cells
XL10-Gold ultracompetent cells transform large plasmids and ligated DNA with highest efficiency.
  • Engineered to transform large plasmids and ligated DNA more efficiently than other cell lines
  • Includes Hte phenotype for 20-fold to 30-fold higher transformation efficiency of ligated DNA and better competent cell performance
  • Chemically competent cells that allow efficient transformation of large DNA molecules, including expression vectors and genomic DNA
  • Ideal for plasmid library construction by decreasing bias against larger plasmids and producing more
More Product Information