GDNF Rapid ELISA Kit: Mouse, Biosensis®
Supplier: Biosensis
BEK-2229-1P BEK-2229-2P
75838-854KT 691.8 CAD
75838-854 75838-856
GDNF Rapid ELISA Kit: Mouse, Biosensis®
Assays ELISAs
The Biosensis GDNF RapidTM enzyme-linked immunosorbent assay (ELISA) Kit is a sandwich ELISA that allows the quantification of GDNF in less than 4 hours in cell culture supernatants, cell lysates, and serum only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application, in particular mouse serum samples.
This ELISA kit consists of a pre-coated anti-GDNF capture antibody, a biotinylated anti-GDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’- tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of GDNF present in samples and protein standards.
This GDNF ELISA kit employs a recombinant mouse GDNF standard and is therefore designed to measure the murine form of GDNF. Note that the antibodies used in this kit cross-react with rat, human and guinea pig GDNF. This kit has not been tested for other applications.
The kit consists of a pre-coated anti-GDNF capture antibody, a biotinylated anti-GDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a coloured reaction product which is directly proportional to the concentration of GDNF present in samples and protein standards.
Delivery information: The ELISA kit box contains 1 or 2×96-well pre-coated strip plate, protein standards, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Caution: It has been configured for research use only and is not to be used in diagnostic or clinical procedures.