GDNF Rapid ELISA Kit: Human, Biosensis®

Supplier: Biosensis

BEK-2222-1P BEK-2222-2P
75838-792KT 691.8 CAD
75838-792 75838-846
GDNF Rapid ELISA Kit: Human, Biosensis®
Assays ELISAs

The Biosensis GDNF RapidTM enzyme-linked immunosorbent assay (ELISA) Kit is a sandwich ELISA that allows the quantification of GDNF in less than 4 hours in cell culture supernatants, cell lysates, and guinea pig serum only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application.


This ELISA kit consists of a pre-coated anti-GDNF capture antibody, a biotinylated anti-GDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’- tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of GDNF present in samples and protein standards.


This GDNF ELISA kit employs a recombinant human GDNF standard obtained from the National Institute for Biological Standards and Control (NIBSC) and is therefore designed to accurately measure the human form of GDNF. Note that the antibodies used in this kit cross-react with guinea pig, rat and mouse GDNF. Guinea pig GDNF has been quantified in serum using the Human GDNF RapidTM ELISA kit, since guinea pig GDNF protein shares closest amino acid sequence homology with human GDNF. Protein levels can be expressed as “human GDNF equivalents”.


This kit has not been tested for other applications.


Delivery information: The ELISA kit box contains 1 or 2 x 96-well pre-coated strip plate, protein standards, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.


Caution: It has been configured for research use only and is not to be used in diagnostic or clinical procedures.

Order Now

SPECIFICATIONS

Environmentally Preferable
Primary antibody reactivity Human
Assay Type Sandwich
Host
Sensitivity Typical limit of detection (LOD) for rhGDNF is <5 pg/mL determined as blank value plus 3x standard deviation of blank OD (n=10)
Detection range 7.8 -500 pg/ml
Cross reactivity The antibodies used in this kit detect guinea pig, mouse and rat GDNF. No cross-reactivity was observed for the following proteins tested at 25 ng/ml in assay buffer: brain-derived neurotrophic factor (rhBDNF), nerve growth factor (rhNGF), neurotrophin-3 (rhNT-3), rhNT-4/5, vascular endothelial growth factor (rhVEGF), recombinant human Neurturin, Artemin and Persephin

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR