SCSI Supercompetent Cells, Agilent Technologies
Supplier: Agilent
200231 200231
CA99900-332EA 333.72 CAD
CA99900-332 99900-332
SCSI Supercompetent Cells, Agilent Technologies
Competent Cells
SCSI supercompetent and DH5α competent cells are classic E. coli strains engineered for competence.
  • Engineered to become competent
This collection of competent cells, including SCSI supercompetent cells and DH5α competent cells, consists of classic E. coli strains that have been engineered to become competent for use in cloning experiments.
Caution: For research use only. Not for use in diagnostic procedures.