LB High-Temperature Bottom Loading Furnace up to 1650 °C, Nabertherm
Supplier: Nabertherm
Nabertherm
LHB157WN3-240 LHB257WN4-240 LHB157WN3-208 LHB157WN3-220 LHB257WN4-208
76538-628EA 28344.17 CAD
76538-628 76538-632 76538-268 76538-630 76538-266
LB High-Temperature Bottom Loading Furnace up to 1650 °C, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
Due to their maximum temperature of 1650 °C and their large chamber, these high-temperature bottom loading furnaces are perfectly suited for sintering translucent zirconia. The motor-driven lifting table significantly simplifies the charging of the high-temperature furnace. The all-around heating of the cylindrical furnace chamber ensures a very even temperature uniformity.
  • Appealing, double-walled stainless steel housing
  • Outstanding temperature uniformity due to all-round furnace chamber heating
  • Exhaust air vent in the roof
  • Type S thermocouple
  • Controller P480 with memory for 50 programs, two sample programs
  • Usable for sintering blanks of all leading manufacturers
  • Drying function: When starting the program the table will be driven in drying position and closes automatically at 500 °C
Tmax 1650 °C furnace chamber with a volume of 1 or 2 liters, table with large floor space contain high-quality heating elements made of molybdenum disilicide offer best possible protection against chemical interaction between charge and heating elements. Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2.
Delivery includes a starter set for charging on one level, additional levels as additional equipment. Precise, motorized toothed belt drive of the table with button operation. Residual drying function for better ventilation of the oven when heating up and also forced cooling function with automatic, step-by-step opening from a preset temperature.
NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive. Equipped with special heating elements made of molybdenum disilicide, chemical interactions between the charge and the furnace components are optimally avoided. The sintered material is placed in saggars made of technical ceramics. Up to two batch containers for max. 15 single crowns per level can be accommodated in the LHT 01/17 LB Speed. The LHT 02/17 LB Speed offers space for up to three saggars for max. 25 individual crowns per level and thus guarantees high productivity.
The high-temperature bottom loading furnaces are additionally equipped with a drying as well as a forced cooling function. For residual drying, the oven remains open gapwise during heating up to a defined temperature and thus ensures reliable removal of moisture. For accelerated cooling, the furnace is automatically opened step by step under program control. Depending on the batch used and the saggars, these high-temperature furnaces can achieve total cycle times of less than two and a half hours. The furnaces can be individually programmed for all recommended sintering curves of all zirconia manufacturers.
Accessories information: Optional adders include: Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; stackable saggars for loading in up to two or three levels, depending on model; process control and documentation via VCD software package for monitoring, documentation and control.
Ordering information: These high-temperature lift-bottom furnaces can be ordered in a variety of voltage options. Models are rated up to 1650 °C. Control with P480 controller (50 programs with 40 segments each) for complex sintering cycles.