AlphaTec® 58-535B Gloves, Ansell
Supplier: Ansell Canada
58535B070 58535B080 58535B090 58535B100 58535B110
CA76268-878CS 849.71 CAD
CA76268-878 CA76268-880 CA76268-882 CA76268-900 CA76268-902
AlphaTec® 58-535B Gloves, Ansell
Gloves
A unique combination of liquid-proof chemical resistance and grip, together with flexibility and dexterity, makes this glove the logical choice when working with chemicals.
  • Medium duty
  • Reliable chemical protection
  • Silicone-free
  • Food safe
The AlphaTec® glove incorporates revolutionary ANSELL GRIP™ technology that enables users to handle wet or oily objects with less grip force and more control. The patented ANSELL GRIP™ technology ensures that the coating polymer does not penetrate the liner during manufacture. The coating is therefore consistent andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Disposable / reusable Reusable
Latex/Latex-free Latex-free
Liner material Acrylic
Finish Ansell Grip™ Technology
Cuff style Gauntlet
Anti-static +
Food contact approved +