Brilliant III SYBR® MM, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
600882 600883 600886 600887
CA97066-528EA 480.77 CAD
CA97066-528 CA97066-530 CA97066-536 CA97066-538
Brilliant III SYBR® MM, Agilent Technologies
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Brilliant III Ultra-Fast SYBR® QPCR and QRT-PCR Master Mixes deliver superior specificity by utilizing a novel faster-activating hot start method to minimize the formation of primer-dimers and other off-target reactions.
  • Reliable and reproducible data
  • Novel fast Taq mutant for qPCR results in under 35 minutes
  • Enhanced rapid hot start capability saves time and reduces primer-dimer formation
  • Convenient pre-blended formulations compatible with any SYBR Green dye
  • Designed for researchers who want access to their data faster without compromising data quality
These unique reagents are equally robust and reproducible across a variety of assays on fast cycling real-time platforms.
Caution: For research use only. Not for use in diagnostic procedures.