dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
Supplier: DLAB SCIENTIFIC INC.
7016201001 7016201002 7016201003 7016201004
76271-358EA 996.17 CAD
76271-358 76271-360 76271-362 76271-364
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
Pipettes Micropipettes
High accuracy, high performance stepper motor ensures accuracy and repeatability, eliminating manual pipetting errors.
  • Two buttons handle all operational settings
  • Lightweight, ergonomic design
  • Small body dimensions enable easy handling and fatigue-free pipetting
  • Adjustable speeds for aspiration and dispensing
  • Li-ion battery and dual charging modes enable longer operation time
  • Self calibration via USB to PC; third party calibration is not required
A motor driven digital control pipette with multifunctional capabilities of pipetting, mixing, stepper, and dilution.
Electrical: 100 to 220 V, 50/60 Hz
Certifications: CE marked. cTUVus.
Delivery information: Each package includes the electronic pipette with battery, stand, charger, USB cable, calibration card, and user manual.