BriskHeat® Full Coverage Insulated Drum Heaters, Cole Parmer
Supplier: Antyila Scientific
09300-50 09300-60
76266-252EA 2533.81 CAD
76266-252 76266-254
BriskHeat® Full Coverage Insulated Drum Heaters, Cole Parmer
Heaters Heating Mantles
Full Coverage Insulated Drum Heaters provide safe, uniform heat to 55 gallon metal drums.
  • Built-in adjustable thermoswitch prevents temperature runaway
  • Lightweight flexible design makes installation quick and easy
Insulated drum heaters provide an efficient method of temperature and viscosity control for liquids. Simply wrap the insulated drum heater around a 55 gallon drum, adjust the built-in thermostat to a desired temperature setting, and uniform heat is evenly distributed around the entire drum.
Certifications: CE approved.