Anti-VEGF Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P275BT-25UG 500-P275BT-50UG
10777-460EA 536.88 CAD
10777-460 10777-462 10777-464
Anti-VEGF Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat VEGF. Anti­Rat VEGF­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR