Anti-RANTES (CCL5) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P78BT-25UG 500-P78BT-50UG
10781-534EA 560.17 CAD
10781-534 10781-536 10781-538
Anti-RANTES (CCL5) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat RANTES (CCL5). Anti­Rat RANTES (CCL5)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: RANTES (CCL5)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR