Anti-IL-2 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P274BT-25UG 500-P274BT-50UG
10777-448EA 536.88 CAD
10777-448 10777-450 10777-452
Anti-IL-2 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-2. Anti­Rat IL-2­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR