Anti-MIP-1beta (CCL4) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P213BT-25UG 500-P213BT-50UG
10776-936EA 536.88 CAD
10776-936 10776-938 10776-940
Anti-MIP-1beta (CCL4) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-1β (CCL4). Anti­Murine MIP-1β (CCL4)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: MIP-1beta (CCL4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR