Anti-MCP-5 (CCL12) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P61BT-25UG 500-P61BT-50UG
10775-520EA 536.88 CAD
10775-520 10781-512 10781-514
Anti-MCP-5 (CCL12) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine MCP-5 (CCL12). Anti­Murine MCP-5 (CCL12)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: MCP-5 (CCL12)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

SPECIFICATIONS

Antigen symbol MCP-5 (CCL12)
Antigen name MCP-5
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Biotin
Reactivity Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Recombinant Murine MCP-5 (CCL12)
Form Lyophilized
Molecular weight 9.3 kDa
Purification Recombinant Murine MCP-5 (CCL12)
Storage buffer See Product Data Sheet for more information.
Storage temperature Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR