Anti-LIX (CXCL6) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P146BT-25UG 500-P146BT-50UG
10771-422EA 560.17 CAD
10771-422 10771-424 10771-426
Anti-LIX (CXCL6) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine LIX (CXCL6). Anti­Murine LIX (CXCL6)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: LIX (CXCL6)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR