Anti-KC (CXCL1) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P115BT-25UG 500-P115BT-50UG
10776-076EA 581.63 CAD
10776-076 10776-078 10776-080
Anti-KC (CXCL1) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine KC (CXCL1). Anti­Murine KC (CXCL1)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: KC (CXCL1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR