Anti-IL-7 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P57BT-25UG 500-P57BT-50UG
10775-484EA 536.88 CAD
10775-484 10775-486 10775-488
Anti-IL-7 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-7. Anti­Murine IL-7­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-7
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR