Anti-IL-1alpha Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P51ABT-25UG 500-P51ABT-50UG
10775-440EA 536.88 CAD
10775-440 10775-442 10775-444
Anti-IL-1alpha Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-1α. Anti­Murine IL-1α­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-1alpha
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR