Anti-C10 (CCL6) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P112BT-25UG 500-P112BT-50UG
10776-040EA 581.63 CAD
10776-040 10776-042 10776-044
Anti-C10 (CCL6) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine C10 (CCL6). Anti­Murine C10 (CCL6)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: C10 (CCL6)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR