Anti-MIP-1alpha (CCL3) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P121BT-25UG 500-P121BT-50UG
10776-148EA 536.88 CAD
10776-148 10776-150
Anti-MIP-1alpha (CCL3) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-1α (CCL3). Anti­Murine MIP-1α (CCL3)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: MIP-1alpha (CCL3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR