Anti-IL-13 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P178BT-25UG 500-P178BT-50UG
10776-664EA 571.15 CAD
10776-664 10776-666
Anti-IL-13 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-13. Anti­Murine IL-13­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-13
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR