Anti-SDF-1alpha (CXCL12) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P87ABT-25UG 500-P87ABT-50UG
10775-804EA 536.88 CAD
10775-804 10775-806 10775-808
Anti-SDF-1alpha (CXCL12) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human SDF-1α (CXCL12). Anti­Human SDF-1α (CXCL12)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: SDF-1alpha (CXCL12)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR