Anti-Persephin Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P138BT-25UG 500-P138BT-50UG
10776-324EA 536.88 CAD
10776-324 10776-326 10776-328
Anti-Persephin Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Persephin. Anti­Human Persephin­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: Persephin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR