Anti-MIP-4 (CCL18) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P108BT-25UG 500-P108BT-50UG
10771-366EA 581.63 CAD
10771-366 10771-368 10771-370
Anti-MIP-4 (CCL18) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human MIP-4 (CCL18). Anti­Human MIP-4 (CCL18)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: MIP-4 (CCL18)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR