Anti-IP-10 (CXCL10) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P93BT-25UG 500-P93BT-50UG
10775-888EA 536.88 CAD
10775-888 10775-890 10775-892
Anti-IP-10 (CXCL10) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IP-10 (CXCL10). Anti­Human IP-10 (CXCL10)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IP-10 (CXCL10)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR