Anti-IL-17D Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P88BT-25UG 500-P88BT-50UG
10775-828EA 536.88 CAD
10775-828 10775-830 10775-832
Anti-IL-17D Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17D. Anti­Human IL-17D­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-17D
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR