Anti-IL-15 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P15BT-25UG 500-P15BT-50UG
10775-052EA 536.88 CAD
10775-052 10775-054 10775-056
Anti-IL-15 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-15. Anti­Human IL-15­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-15
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR