Anti-IL-11 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P01BT-25UG 500-P01BT-50UG
10774-940EA 536.88 CAD
10774-940 10774-942 10774-944
Anti-IL-11 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-11. Anti­Human IL-11­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-11
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR