Anti-Galectin-3 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P246BT-25UG 500-P246BT-50UG
10777-212EA 536.88 CAD
10777-212 10777-214 10777-216
Anti-Galectin-3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Galectin-3. Anti­Human Galectin-3­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: Galectin-3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR