Anti-FGF-acidic Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P17BT-25UG 500-P17BT-50UG
10775-076EA 536.88 CAD
10775-076 10775-078 10775-080
Anti-FGF-acidic Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-acidic. Anti­Human FGF-acidic­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: FGF-acidic
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol FGF-acidic
Antigen name FGF-acidic
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Biotin
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Recombinant Human FGF-acidic
Form Lyophilized
Molecular weight 16.8 kDa
Purification Recombinant Human FGF-acidic
Storage buffer See Product Data Sheet for more information.
Storage temperature Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR