Anti-CTGFL/WISP-2 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P212BT-25UG 500-P212BT-50UG
10776-924EA 536.88 CAD
10776-924 10776-926 10776-928
Anti-CTGFL/WISP-2 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human CTGFL/WISP-2. Anti­Human CTGFL/WISP-2­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: CTGFL/WISP-2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR