Anti-VEGF Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P275-50UG 500-P275-100UG 500-P275-1MG
10777-454EA 536.88 CAD
10777-454 10777-456 10777-458
Anti-VEGF Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat VEGF. Anti­Rat VEGF­specific antibody was purified by affinity chromatography employing an immobilized Rat VEGF165 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR