Anti-SCF Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P202-50UG 500-P202-100UG 500-P202-1MG
10776-854EA 536.88 CAD
10776-854 10776-856 10776-858
Anti-SCF Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat SCF. Anti­Rat SCF­specific antibody was purified by affinity chromatography employing an immobilized Rat SCF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: SCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR