Anti-IP-10 (CXCL10) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P290-50UG 500-P290-100UG 500-P290-1MG
10777-510EA 536.88 CAD
10777-510 10777-512 10777-514
Anti-IP-10 (CXCL10) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat IP-10 (CXCL10). Anti­Rat IP-10 (CXCL10)­specific antibody was purified by affinity chromatography employing an immobilized Rat IP-10 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IP-10 (CXCL10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR