Anti-IL-1beta Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P80-50UG 500-P80-100UG 500-P80-1MG
10775-714EA 560.17 CAD
10775-714 10775-716 10775-718
Anti-IL-1beta Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-1β. Anti­Rat IL-1β­specific antibody was purified by affinity chromatography employing an immobilized Rat IL-1β matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-1beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR