Anti-MIP-1beta (CCL4) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P213-50UG 500-P213-100UG 500-P213-1MG
10776-930EA 536.88 CAD
10776-930 10776-932 10776-934
Anti-MIP-1beta (CCL4) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-1β (CCL4). Anti­Murine MIP-1β (CCL4)­specific antibody was purified by affinity chromatography employing an immobilized Murine MIP-1β matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MIP-1beta (CCL4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR