Anti-MIP-1alpha (CCL3) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P121-50UG 500-P121-100UG 500-P121-1MG
10776-142EA 536.88 CAD
10776-142 10776-144 10776-146
Anti-MIP-1alpha (CCL3) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-1α (CCL3). Anti­Murine MIP-1α (CCL3)­specific antibody was purified by affinity chromatography employing an immobilized Murine MIP-1α matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MIP-1alpha (CCL3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR