Anti-MCP-5 (CCL12) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P61-50UG 500-P61-100UG 500-P61-1MG
10775-514EA 560.17 CAD
10775-514 10775-516 10775-518
Anti-MCP-5 (CCL12) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine MCP-5 (CCL12). Anti­Murine MCP-5 (CCL12)­specific antibody was purified by affinity chromatography employing an immobilized Murine MCP-5 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MCP-5 (CCL12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR