Anti-IL-7 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P57-50UG 500-P57-100UG 500-P57-1MG
10775-478EA 536.88 CAD
10775-478 10775-480 10775-482
Anti-IL-7 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-7. Anti­Murine IL-7­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-7 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR