Anti-IL-4 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P54BT-50UG 500-P54BT-25UG
10771-312EA 686.72 CAD
10771-312 10771-314 10775-464
Anti-IL-4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-4. Anti­Murine IL-4­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR