Anti-IL-3 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P53-50UG 500-P53-100UG 500-P53-1MG
10775-446EA 536.88 CAD
10775-446 10775-448 10775-450
Anti-IL-3 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-3. Anti­Murine IL-3­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-3 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR