Anti-IL-22 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P223-50UG 500-P223-100UG 500-P223-1MG
10776-990EA 536.88 CAD
10776-990 10776-992 10776-994
Anti-IL-22 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-22. Anti­Murine IL-22­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-22 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR