Anti-IL-21 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P278-50UG 500-P278-100UG 500-P278-1MG
10771-520EA 560.17 CAD
10771-520 10771-522 10771-524
Anti-IL-21 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-21. Anti­Murine IL-21­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-21 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR